ENAH03856596
Enamine
(5-methyl-1,2-oxazol-3-yl)methanethiol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(5-methylisoxazol-3-yl)methanethiol
Inchi Code
1S/C5H7NOS/c1-4-2-5(3-8)6-7-4/h2,8H,3H2,1H3
InChI key
INDASJJIKPECHA-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-746792
MSDS
https://www.enaminestore.com/catalog/EN300-746792
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service