ENA465562124
Enamine
methyl 3-(chlorosulfonyl)pyridine-2-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 3-(chlorosulfonyl)-2-pyridinecarboxylate
Inchi Code
1S/C7H6ClNO4S/c1-13-7(10)6-5(14(8,11)12)3-2-4-9-6/h2-4H,1H3
InChI key
NPGWMJZYBJTPRV-UHFFFAOYSA-N
storage temp.
4
Purity
95
Country of Origin
UA
Shipping Temperature
Ice Pack
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-218017
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-218017
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.