Skip to Content
Merck
All Photos(1)

Key Documents

CIAH987EDFC6

ChemScene LLC (Preferred Partner)

Deschlorodemethyldiazepam

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
236.27
MDL number:

bp

424.4 C at 760 mmHg

mp

170-180 C

Iupac Name

5-phenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-one

Inchi Code

1S/C15H12N2O/c18-14-10-16-15(11-6-2-1-3-7-11)12-8-4-5-9-13(12)17-14/h1-9H,10H2,(H,17,18)

InChI key

IVUAAOBNUNMJQC-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/2898-08-0.html

Boiling point

424.4 C at 760 mmHg

Melting Point

170-180 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-D0048.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library