Skip to Content
Merck
All Photos(1)

Key Documents

CH6447917138

Chem Impex International, Inc.

Tris-succinimidyl-1,3,5-benzenetricarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
501.36
MDL number:

Pricing and availability is not currently available.

mp

247-251 C

Iupac Name

1-[(3,5-bis{[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl}benzoyl)oxy]-2,5-pyrrolidinedione

Inchi Code

1S/C21H15N3O12/c25-13-1-2-14(26)22(13)34-19(31)10-7-11(20(32)35-23-15(27)3-4-16(23)28)9-12(8-10)21(33)36-24-17(29)5-6-18(24)30/h7-9H,1-6H2

InChI key

BCFNKSPKXMSVLG-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

99%-6..

Shipping Temperature

RT

Physical Form

White solid

Product Link

https://www.chemimpex.com/product/productinfo/tris-succinimidyl-1-3-5-benzenetricarboxylate/13656?cid=0

Melting Point

247-251 C

MSDS

https://www.chemimpex.com/product/productinfo/tris-succinimidyl-1-3-5-benzenetricarboxylate/13656?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library