C12H93E354F8
Chemcia Scientific, LLC
2-Trifluoromethyl-imidazo[1,2-a]pyridine-6-carbaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-(trifluoromethyl)imidazo[1,2-a]pyridine-6-carbaldehyde
Inchi Code
1S/C9H5F3N2O/c10-9(11,12)7-4-14-3-6(5-15)1-2-8(14)13-7/h1-5H
InChI key
FVMJCPVTPRZEON-UHFFFAOYSA-N
storage temp.
0-5C
Purity
95%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Light Yellow Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=11459
MSDS
http://www.chemcia.com/product_fl02.asp?id=11459
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.