Skip to Content
Merck
All Photos(1)

Key Documents

C12H316CE805

Chemcia Scientific, LLC

7-Bromo-2,6-dimethyl-thieno[3,2-d]pyrimidin-4-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
258.14
MDL number:

Iupac Name

7-bromo-2,6-dimethylthieno[3,2-d]pyrimidin-4-ylamine

Inchi Code

1S/C8H8BrN3S/c1-3-5(9)6-7(13-3)8(10)12-4(2)11-6/h1-2H3,(H2,10,11,12)

InChI key

ZDPDZMBXDAMDFZ-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=363

MSDS

http://www.chemcia.com/product_fl02.asp?id=363


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library