Skip to Content
Merck
All Photos(1)

Key Documents

D-070

Supelco

N-Desmethylolanzapine-d8 dihydrochloride solution

100 μg/mL in acetonitrile: water (1:1) (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

N-Desmethylolanzapine-D8 dihydrochloride solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
1.000,00 kr.

1.000,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
1.000,00 kr.

About This Item

Empirical Formula (Hill Notation):
C16D8H10N4S · H2Cl2
Molecular Weight:
379.38
UNSPSC Code:
41116107
NACRES:
NA.24

1.000,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile: water (1:1) (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

General description

A stable-labeled internal standard for use in LC/MS or GC/MS applications including clinical toxicology analysis, therapeutic drug monitoring, or isotope dilution methods. Desmethylolanzapine is the primary metabolite of olanzapine, marketed as the atypical antipsychotic Zyprexa® for the treatment of schizophrenia and bipolar disorder.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Zyprexa is a registered trademark of Eli Lilly and Co.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service