Skip to Content
Merck
All Photos(1)

Key Documents

878116P

Avanti

C18-04:0 PC

Avanti Research - A Croda Brand 878116P, powder

Synonym(s):

1-0-octadecyl-2-butyryl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H62NO7P
CAS Number:
Molecular Weight:
579.79
UNSPSC Code:
12352211

form

powder

packaging

pkg of 1 × 5 mg (878116P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878116P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCC)=O)COCCCCCCCCCCCCCCCCCC)=O

General description

C18-04:0 PC, also called as 1-O-octadecyl-2-butyryl-sn-glycero-3-phosphocholine, belongs to a class of bioactive lipids. Its structure contains ether-linked octadecyl chain at the sn-1 position and a butyryl group at the sn-2 position.

Packaging

5 mL Clear Glass Sealed Ampule (878116P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunil A Nankar et al.
Biochimie, 95(6), 1196-1207 (2013-02-05)
Apolipoprotein-derived peptides are promising candidates for the treatment of various inflammatory conditions and the main mechanism proposed for the protective action of these peptides includes binding to pro-inflammatory lipid mediators with high affinity and facilitating their sequestration/metabolism/clearance in the body.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service