Skip to Content
Merck
All Photos(1)

Documents

860619P

Avanti

CER6-2′S(d9)

Avanti Research - A Croda Brand 860619P, powder

Synonym(s):

N-(2′-(S)-hydroxypalmitoyl(d9)) D-ribo-phytosphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H60D9NO5
CAS Number:
Molecular Weight:
580.97
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860619P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860619P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@]([H])(CCCCCCCCCCCCCC)[C@](O)([H])[C@@]([H])(NC([C@@H](O)CCCCCCCCCCC([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])=O)CO

General description

CER6-2′S(d9), also known as N-(2′-(S)-hydroxypalmitoyl(d9)) D-ribo-phytosphingosine is a deuterium-labeled skin ceramide standard.

Packaging

5 mL Amber Glass Screw Cap Vial (860619P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service