Skip to Content
Merck
All Photos(1)

Documents

860565P

Avanti

18:1-6:0 Biotin PI(3,5)P2

Avanti Research - A Croda Brand 860565P, powder

Synonym(s):

1-oleoyl-2-[6-biotinyl(aminohexanoyl)]-sn-glycero-3-phosphoinositol-3,5-bisphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H87N6O21P3S
CAS Number:
Molecular Weight:
1149.16
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (860565P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (860565P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 860565P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O[C@H]1[C@H](O)[C@@H](OP([O-])(O)=O)[C@H](O)[C@@H](OP(O)([O-])=O)[C@H]1O)([O-])=O)(OC(CCCCCNC(CCCC[C@]2([H])SC[C@@]3([H])[C@]2([H])NC(N3)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+].[NH4+].[NH4+]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (860565P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (860565P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service