Skip to Content
Merck
All Photos(2)

Key Documents

860409C

Avanti

9-PAHSA-d9

Avanti Research - A Croda Brand 860409C

Synonym(s):

9-[((13,13,14,14,15,15,16,16,16-d9)palmitoyl)hydroxy]-stearic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
3.500,00 kr.

3.500,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
3.500,00 kr.

About This Item

Empirical Formula (Hill Notation):
C34H57D9O4
CAS Number:
Molecular Weight:
547.94
UNSPSC Code:
12352211
NACRES:
NA.25

3.500,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (860409C-1mg)
pkg of 1 × 1 mL (860409C-500ug)

manufacturer/tradename

Avanti Research - A Croda Brand 860409C

concentration

0.5 mg/mL (860409C-500ug)
1 mg/mL (860409C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCC(OC(CCCCCCCCCCCC([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])=O)CCCCCCCC(O)=O

InChI key

MHQWHZLXDBVXML-NPNXCVMYSA-N

General description

9-PAHSA-d9 9-[((13,13,14,14,15,15,16,16,16-d9)palmitoyl)hydroxy]-stearic acid is a deuterated palmitoyl hydroxy-stearic acid wherein the nine protons of palmitoyl are replaced by deuterium. Palmitic acid-9-hydroxy-stearic acid (9-PAHSA) is the most abundantly found fatty acid ester of hydroxy fatty acids (FAHFA).[1]
PAHSA levels correlate highly with insulin sensitivity and are reduced in adipose tissue and serum of insulin-resistant humans. In adipocytes, PAHSAs signal through GPR120 to enhance insulin-stimulated glucose uptake. Thus, FAHFAs are endogenous lipids with the potential to treat type 2 diabetes. Avanti has obtained a license from the patent holder to manufacture these products for research use.

Packaging

5 mL Clear Glass Sealed Ampule (860409C-1mg)
5 mL Clear Glass Sealed Ampule (860409C-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of lysohematoside from hematoside of equine erythrocyte and its chemical and hemolytic properties.
T Taketomi et al.
Journal of biochemistry, 68(4), 475-485 (1970-10-01)
Elsa Pflimlin et al.
Cell metabolism, 28(2), 217-227 (2018-06-26)
Fatty acid esters of hydroxylated fatty acids (FAHFAs) were discovered as a novel class of endogenous mammalian lipids whose profound effects on metabolism have been shown. In the current study, in vitro and in vivo the metabolic effects of two of these
Matthew J Kolar et al.
Biochemistry, 55(33), 4636-4641 (2016-08-11)
A recently discovered class of endogenous mammalian lipids, branched fatty acid esters of hydroxy fatty acids (FAHFAs), possesses anti-diabetic and anti-inflammatory activities. Here, we identified and validated carboxyl ester lipase (CEL), a pancreatic enzyme hydrolyzing cholesteryl esters and other dietary
Mark M Yore et al.
Cell, 159(2), 318-332 (2014-10-11)
Increased adipose tissue lipogenesis is associated with enhanced insulin sensitivity. Mice overexpressing the Glut4 glucose transporter in adipocytes have elevated lipogenesis and increased glucose tolerance despite being obese with elevated circulating fatty acids. Lipidomic analysis of adipose tissue revealed the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service