Skip to Content
Merck
All Photos(2)

Key Documents

850851P

Avanti

16:0 Monomethyl PE

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-methyl, powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N-methyl ; 110743

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
1.790,00 kr.

1.790,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
1.790,00 kr.

About This Item

Empirical Formula (Hill Notation):
C38H76NO8P
CAS Number:
Molecular Weight:
705.99
UNSPSC Code:
51191904
NACRES:
NA.25

1.790,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850851P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850851P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)([H])[H])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C38H76NO8P/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-37(40)44-34-36(35-46-48(42,43)45-33-32-39-3)47-38(41)31-29-27-25-23-21-19-17-15-13-11-9-7-5-2/h36,39H,4-35H2,1-3H3,(H,42,43)

InChI key

QSBINWBNXWAVAK-UHFFFAOYSA-N

General description

16:0 Monomethyl phosphoethanolamine (PE) is an alkyl phosphate derivative.

Packaging

5 mL Amber Glass Screw Cap Vial (850851P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service