Skip to Content
Merck
All Photos(2)

Key Documents

850528P

Avanti

Mal(11.2)

tridecan-3-yloxy-β-D-maltoside, powder

Synonym(s):

3-tridecyl-Β-D-maltopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
2.330,00 kr.

2.330,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
2.330,00 kr.

About This Item

Empirical Formula (Hill Notation):
C25H48O11
CAS Number:
Molecular Weight:
524.64
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

2.330,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850528P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand 850528P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OC(CC)CCCCCCCCCC)[C@H](O)[C@@H](O)[C@@H]1O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2

InChI key

CAAOONAGUQWVFE-MXEARUNBSA-N

General description

This new class of detergents contain a short, branched alkyl chain at the interface between the polar head and the apolar tail. This design mimics the second aliphatic chain of lipid molecules and reduces water penetration, thereby increasing the hydrophobicity within the interior of the micelle. A detergent architecture of this style allows formation of smaller, more compact micelles which provides improved performance in the stabilization and crystallization of integral membrane proteins.

Packaging

20 mL Clear Glass Screw Cap Vial (850528P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxu Jiang et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(12), E698-E704 (2012-02-23)
LacY mutant Cys154 → Gly exhibits a periplasmic-closed crystal structure identical to the WT, but is periplasmic-open in the membrane. The mutant hardly catalyzes transport, but binds galactosides from either side of the membrane with the same affinity and is
Wen-Xu Hong et al.
Langmuir : the ACS journal of surfaces and colloids, 26(11), 8690-8696 (2010-03-18)
A challenging requirement for structural studies of integral membrane proteins (IMPs) is the use of amphiphiles that replicate the hydrophobic environment of membranes. Progress has been impeded by the limited number of useful detergents and the need for a deeper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service