Skip to Content
Merck
All Photos(1)

Documents

850189P

Avanti

18:0-20:4 PI(3)P

powder

Synonym(s):

1-stearoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′-phosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H90N2O16P2
CAS Number:
Molecular Weight:
1001.17
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

99% (TLC)

form

powder

packaging

package of 1 × 100 μg (850189P-100UG)
package of 1 × 500 μg (850189P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](OP(O)([O-])=O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

General description

Phosphatidylinositol 3‐phosphate (PI(3)P) is obtained from the phosphorylation of inositol ring of phosphatidylinositol. PI(3)P is localized in the early endosomes, multivesicular endosomes and autophagosomes.

Biochem/physiol Actions

Phosphatidylinositol 3‐phosphate (PI(3)P) regulates the autophagy in lysosomes.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850189P-100UG)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850189P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylinositol 3-phosphates?at the interface between cell signalling and membrane traffic
Marat A L and Haucke V
The Embo Journal, 35(6), 561-579 (2016)
Phosphatidylinositol-3-phosphate in the regulation of autophagy membrane dynamics
Nascimbeni A C, et al.
FEBS Journal, 284(9), 1267-1278 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service