Skip to Content
Merck
All Photos(1)

Documents

840538L

Avanti

18:1 MGDG(d18)

Avanti Research - A Croda Brand

Synonym(s):

18:1 MGDG(d18), 1,2-Dioleoyl-3-β-D-galactopyranosyl-sn-glycerol (d18), monogalactosyldioleoylglycerol (d18)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H64D18O10
Molecular Weight:
801.24
UNSPSC Code:
12352211
NACRES:
NA.21

Assay

>99% (TLC)

form

(DCM:Methanol (1:1) solution)

packaging

pkg of 1 × 1 mg (840538L-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

shipped in

dry ice

storage temp.

−20°C

Application

lipidomics

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service