Skip to Content
Merck
All Photos(1)

Key Documents

810222P

Avanti

C6-NBD Glucosyl Ceramide

Avanti Research - A Croda Brand 810222P, powder

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-D-glucosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

810222P-1MG
6.500,00 kr.
250 μG
10.510,00 kr.

6.500,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
810222P-1MG
6.500,00 kr.
250 μG
10.510,00 kr.

About This Item

Empirical Formula (Hill Notation):
C36H59N5O11
CAS Number:
Molecular Weight:
737.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

6.500,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810222P-1MG)
pkg of 1 × 250 μg (810222P-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810222P

shipped in

dry ice

storage temp.

−20°C

Biochem/physiol Actions

The synthesis of glucosyl ceramide (GlcCer) occurs in the cytosolic membranes of Golgi. It is catalysed by GlcCer synthase. In the luminal Golgi membrane, GlcCer participates in the generation of glycosphingolipids.[1] C6-NBD glcCer is useful to view ceramide rich membrane domains in Golgi compartments.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810299P-1MG)
5 mL Clear Glass Sealed Ampule (810222P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A functionalized sphingolipid analogue for studying redistribution during activation in living T cells
Collenburg L, et al.
Journal of Immunology, 196(9), 3951-3962 (2016)
Glycosphingolipid synthesis requires FAPP2 transfer of glucosylceramide
D?Angelo G, et al.
Nature, 449(7158), 62-62 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service