330379W
Avanti
Carnitine SPLASH™
Avanti Research™ - A Croda Brand
Synonym(s):
Carnitine SPLASH LIPIDOMIX Quantitative Mass Spec Standard
Select a Size
5.910,00 kr.
Select a Size
About This Item
5.910,00 kr.
Recommended Products
biological source
synthetic
description
Technique(s): mass spectrometry
Assay
≥99% (TLC)
packaging
pkg of 1 × 1 ea (330379W-1EA)
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−20°C
General description
Application
Packaging
Legal Information
Kit Components Also Available Separately
- 870311PL-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870333PC2:0 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870332PC3:0 L-carnitine-d9 (HCl Salt), Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870331PC4:0 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870330PC5:0 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870329PC6:0 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870328PC8:0 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870327PC10:0 L-carnitine-d9 (HCl salt), Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870326PC12:0 L-carnitine-d9 (HCl salt), Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870325PC14:0 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870323PC16:0 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870324PC18:0 L-carnitine-d9 (HCl salt), Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870321PC18:1 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
- 870322PC18:2 L-carnitine-d9, Avanti Research™ - A Croda Brand 10µg/mLSDS
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service