Skip to Content
Merck
All Photos(1)

Key Documents

ATEH99BCA11F

AstaTech, Inc.

BOC-ALA-OTBU

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
245.32
MDL number:

Iupac Name

tert-butyl (2S)-2-[(tert-butoxycarbonyl)amino]propanoate

Inchi Code

1S/C12H23NO4/c1-8(9(14)16-11(2,3)4)13-10(15)17-12(5,6)7/h8H,1-7H3,(H,13,15)/t8-/m0/s1

InChI key

QIARYQWAMYNMRX-QMMMGPOBSA-N

Purity

97%

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=142207

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C10225


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library