ATE514320207
AstaTech, Inc.
6-BROMO-2,2-DIMETHYL-4H-BENZO[D][1,3]DIOXINE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
6-bromo-2,2-dimethyl-4H-1,3-benzodioxine
Inchi Code
1S/C10H11BrO2/c1-10(2)12-6-7-5-8(11)3-4-9(7)13-10/h3-5H,6H2,1-2H3
InChI key
UIALGZOSKLCMHF-UHFFFAOYSA-N
storage temp.
2-8 C
Purity
95%
Shipping Temperature
Normal
Physical Form
Colorless to Yellow Liquid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=84751
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=F51269
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.