Skip to Content
Merck
All Photos(1)

Key Documents

APOH93D9D644

Apollo Scientific Ltd

2-(Butane-1-sulfonyl)aniline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
249.76
MDL number:
Pricing and availability is not currently available.

Iupac Name

2-(butylsulfonyl)aniline hydrochloride

Inchi Code

1S/C10H15NO2S.ClH/c1-2-3-8-14(12,13)10-7-5-4-6-9(10)11;/h4-7H,2-3,8,11H2,1H3;1H

InChI key

MRKPWUOGOFDTJM-UHFFFAOYSA-N

Purity

98%

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library