APO455835268
Apollo Scientific Ltd
4-Chloro-2-(trifluoromethyl)benzonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-chloro-2-(trifluoromethyl)benzonitrile
Inchi Code
1S/C8H3ClF3N/c9-6-2-1-5(4-13)7(3-6)8(10,11)12/h1-3H
InChI key
GRNQHTXPUDZMGB-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
98%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/4-chloro-2-trifluoromethylbenzonitrile
Synonym
5-Chloro-2-cyanobenzotrifluoride, 4-Chloro-alpha,alpha,alpha-trifluoro-o-tolunitrile
Flash Point
109/10mm
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/4-chloro-2-trifluoromethylbenzonitrile
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.