Key Documents
Z742586
Accuris M1 Grade Calibration Weight
size 1000 g, stainless steel
Select a Size
1 EA
944,00 kr.
944,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
944,00 kr.
About This Item
UNSPSC Code:
41111500
NACRES:
NB.35
944,00 kr.
Please contact Customer Service for Availability
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific W1005-1000
size
1000 g
General description
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)