Key Documents
Z742328
2MAG maxMIX single point stirrers up to 150 L,
AC/DC input 100-240 V, US 3-pin plug
Synonym(s):
high volume stirrer, induction plate, induction stirrer, laboratory stirrer, magnetic stir plate, magnetic stirrer
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
Product Z742328 is not currently sold in your country. Contact Technical Service
material
stainless steel housing
stainless steel plate
feature
positions 1
40 W Stirring Power (
4 step)
packaging
pack of 1 ea
manufacturer/tradename
2mag 30501
parameter
-10-50 °C temp. limit (at 80 percent humidity)
100-1200 rpm speed
W × D × H
265 mm × 320 mm × 68 mm
plate size × W × L
× 260 mm × 290 mm
stirrer volume
1-150 L , per stirring position
weight
~9.3 kg
General description
Unbeaten strong magnetic stirrer with large set-up area, up to 150 litres stirring volume, also suitable for highly viscous media.
Very strong magnetic coupling, therefore predestined for magnetic power transmission over wide distances (up to 60 mm), for thick vessel bottoms or for double-coat vessels. Maintenance-free 2mag-motor-drive with large speed range from 100 up to 1,200 rpm, fully encapsulated and tight stainless steel housing, IP64, extremely robust construction and performance, easy to clean under running water and firm, individual measurements on request. Integrated control unit with strong 40 watts stirring power,large speed range from 100 up to 1,200 rpm, SoftStart-function (variable 20-99 seconds) for reliable catching/centring and safe accelerating of the stirring bar at different volumes and viscosities.
Choose from one of the most recent versions:
Certificates of Analysis (COA)