Key Documents
Z563137
J-KEM® digital temperature monitor
thermocouple type, K, temp. range -50-1200 °C
About This Item
UNSPSC Code:
41112303
Pricing and availability is not currently available.
AC/DC input
120 V
feature
thermocouple type K
manufacturer/tradename
J-Kem
parameter
-50-1200 °C temp. range
Related Categories
General description
Displays the temperature of an attached reaction or other piece of equipment on a bright LED display. No alarms. 0.1 °C resolution.
Legal Information
J-KEM is a registered trademark of J-KEM Electronics, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)