Z308978
Quickfit® ground sleeve stirrer glands
shaft D 11 mm, joint: ST/NS 24/29
Synonym(s):
ST20/3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS24/29
manufacturer/tradename
Quickfit ST 20/3
availability
not available in USA and Canada
shaft D
11 mm
fitting
24 thread
Looking for similar products? Visit Product Comparison Guide
General description
Shaft that passes through screw thread adapterr in top of gland. Will operate at up to 700rpm and down to 0.1 torr. Cup shaped lubricant reservoir surrounds sleeve such that lubricant also acts as coolant.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service