Skip to Content
Merck
All Photos(1)

Documents

Z272086

NMR tube caps

NMR tube size 8 mm

Synonym(s):

cap tubes, nmr tubes, plastic end cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
SB.32

packaging

pkg of 100 ea

manufacturer/tradename

Wilmad 521B

NMR tube size

8 mm

color

white

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service