Skip to Content
Merck
All Photos(1)

Key Documents

Z250392

Sigma-Aldrich

Organic Solvents: Physical Properties and Methods of Purification, 4th Edition

Sign Into View Organizational & Contract Pricing


About This Item

ISBN-10:
0471084670
ISBN-13:
978-047108467-9
UNSPSC Code:
55101509
Pricing and availability is not currently available.

publication info

J. Riddick, et al., John Wiley & Sons, Ltd, 1986, 1344 pp., hard cover

General description

The fourth edition updates and expands the material of the third edition. Besides updating the physical properties and preparation techniques of previously noted solvents, this edition includes over 150 new solvents, many selected to complete groups of isomers and expand homologous series listed in the third edition. Several isomeric and homologous series are initiated here, including the xylenols, chlorinated toluenes, xylidenes, picolines, lutidines, and silanes. The increasing importance of aerosol propellants is recognized with over a dozen propellants represented among the lighter hydrocarbons, aliphatic fluorinated hydrocarbons, and mixed halogenated hydrocarbons. Numerical values are now presented in cgs and SI units. Six new tabulated properties are included: pressure coefficient dp/dt, isothermal compressibility kT, adiabatic compressibility kS, thermal conductivity ?, solubility parameter d and evaporation rates. In addition, the method of citing journal references in the bibliography has been changed to conform more nearly to the style of Chemical Abstracts.

Table of Contents

Classification of the Solvents.
Physical Properties: Discussion of Properties and Criteria for Their Selection.
Physical Properties: Tabulations.
Criteria of Purity.
Drying and Determination of Water.
Purification Methods.
Bibliography.
Index.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service