Z180122
Ace large chromatography column
I.D. × L 100 mm × 1,200 mm, capacity 9.43 L
About This Item
grade
for chromatography
manufacturer/tradename
Ace Glass 5820116
technique(s)
LPLC: suitable
I.D. × L
100 mm × 1,200 mm
capacity
9.43 L
Looking for similar products? Visit Product Comparison Guide
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service