Key Documents
Z163619
Sigma-Aldrich
MiniPax® absorbent packets
molecular sieves, weight 1 g (desiccant), L × W 3/4 in. × 1 7/16 in.
Synonym(s):
Absorbent Packets
About This Item
UNSPSC Code:
24141516
NACRES:
NB.43
manufacturer/tradename
Multisorb 02-00041AG104
L × W
3/4 in. × 1 7/16 in.
weight
1 g (desiccant)
General description
MiniPax® packets are compact, extremely durable, non-dusting, and non-linting with high water vapor trasmission rate and controlled porosity. The packets take up little space in packaging and have a high moisture adsorbing capacity.
Envelopes are formed from sheet of polyethylene fibers has high wet strength and is permeable to both gases and odors. Indicating MiniPax® contain cobalt chloride and should not be used for food or pharmaceutical applications.
  • Compact - The low profile and compact shape of MiniPax® packets saves space and displaces a minimum amount of product. Its flat contour eliminates any potential confusion with a capsule or a pill
  • Maximum surface exposure - The exceptionally narrow seal skirt on only two ends gives MiniPax® packets an unusually large adsorptive surface area for its overall size
  • Integral window - An integral transparent strip permits observation of the contained adsorbent and allows effective use of indicating desiccants to show when moisture has been adsorbed and desiccant should be replaced
Legal Information
MiniPax is a registered trademark of Multisorb Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)