Skip to Content
Merck
All Photos(4)

Key Documents

WHA77015102

Whatman® UNIPLATE microplates

24 well, round well bottom, well volume 10 mL, natural polypropylene

Synonym(s):

Z746746, micro plate, multiwell plate, well plates

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
3.520,00 kr.

3.520,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
3.520,00 kr.

About This Item

UNSPSC Code:
41122107
NACRES:
NB.24
material:
V-bottom wells
natural polypropylene
size:
24 well
sterility:
non-sterile
feature:
lid: no

3.520,00 kr.


Please contact Customer Service for Availability

material

V-bottom wells
natural polypropylene

sterility

non-sterile

feature

lid: no

packaging

pkg of 25 ea

manufacturer/tradename

Cytiva
Whatman 7701-5102
Whatman Article No. 28420867 (US reference)

technique(s)

PCR: suitable

size

24 well

well volume

10 mL

working volume

10 mL

Looking for similar products? Visit Product Comparison Guide

General description

Whatman offers a wide range of UNIPLATE microplates including various well profiles, well volumes, and well densities, in diverse polymer materials. Most UNIPLATE microplates fit most microplate readers and automated plate handling devices.

Application

  • Sample storage
  • Assay development
  • High-throughput screening
  • Plasmid miniprep
  • ELISA assays
  • Luminescence/chemiluminescence
  • Cell culture
  • Filtrate collection

Features and Benefits

  • Widest selection from a single source: Choice of well volumes ranging from 8NA µl to 1NA ml, well densities from 24 to 384 wells with round or ′V′ bottom for maximum recovery.
  • Chemical compatibility: Available in chemically resistant polymers capable of withstanding low temperatures for long-term storage. Opaque plates prevent optical cross talk in light emitting assays.
  • Conforms to ANSI/SBS microplate standard.
  • Guaranteed for use with robotic handlers and centrifuge carriers.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service