WHA68952504
Whatman® GD/X syringe filters
glass microfiber membrane, pore size 0.45 μm, diam. 25 mm, non-sterile, pk of 1500 ea
Synonym(s):
Whatman filter, Z745529, syringe filter, syringe filters
Select a Size
26.800,00 kr.
Select a Size
About This Item
26.800,00 kr.
Recommended Products
material
glass microfiber membrane
polypropylene housing
sterility
non-sterile
feature
glass fiber prefilter
packaging
pk of 1500 ea
manufacturer/tradename
Whatman 6895-2504
Whatman Article No. 28420547 (US reference)
diam.
25 mm
volume
≤25 mL
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service