Skip to Content
Merck
All Photos(1)

Key Documents

WHA2712T

Whatman® Polycap HD heavy duty encapsulated filter

Polycap HD 75, pore size 1.0-5.0 μm

Synonym(s):

Whatman filter, Whatman filters, Z741876

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
3.600,00 kr.

3.600,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
3.600,00 kr.

About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

3.600,00 kr.


Please contact Customer Service for Availability

material

polypropylene housing
polypropylene membrane

sterility

non-sterile

packaging

pkg of 5 ea

manufacturer/tradename

Whatman 2712T
Whatman Article No. 28418952 (US reference)

diam.

75 mm

pore size

1.0-5.0 μm

fitting

inlet 0.125 inch FNPT
outlet 0.0125 inch FNPT

Looking for similar products? Visit Product Comparison Guide

General description

Whatman Polycap HD capsule filters are heavy duty polypropylene filters, commonly used in processing applications for general sample preparation; fine filtration; and refiltration for reverse osmosis, ultrafiltration and microfiltration membranes.

Heavy duty polypropylene filter capsule for chemical processing

Whatman Polycap HD capsule filters are used in processing applications to filter buffers, solvents, reagents and emulsions across a wide range of industries including clean air and gas equipment, cosmetics and personal care, food and beverage, inks and pigments, pharmaceuticals, photography, semiconductors and magnetic media.

Chemical processing applications often require a capsule filter to ensure a finishing step that yields a high purity final product. Different solution temperatures and pH values can complicate filtration. Whatman Polycap HD capsule filters are manufactured with polypropylene filter media, support system and housing allow use with a broad range of solutions, pH, and temperatures. Sturdy construction enables high flow rate and high retention capacity for excellent filtrate purity.

Features and Benefits

  • Can be autoclaved with steam at 121°C for 20 min
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
  • Available in 0.45, 1.0, 5.0, 10, or 20 µm pore sizes, as well as a variety of end-fitting configurations

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service