WHA10404139
Whatman® cellulose acetate membrane filters 0.2 μm pore size
white, hydrophilic, 25 ea, 293 mm diam
Synonym(s):
Z750352
About This Item
Recommended Products
product name
Whatman® cellulose acetate membranes, OE66 membrane circles, cellulose acetate, pore size 0.2 μm, diam. 293 mm
material
cellulose acetate
cellulose acetate membrane
sterility
non-sterile
packaging
pk of 25
manufacturer/tradename
Whatman 10404139
Whatman Article No. 28415150 (US reference)
diam.
293 mm
pore size
0.2 μm
suitability
suitable for (biological and clinical analysis, sterility tests, and scintillation measurements)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
High chemical resistance membranes
Due to their high chemical resistance, cellulose acetate membrane filters from Cytiva′s business reliably prepare liquid scintillation cocktail for scintillation counter measurement. Additionally, the highly heat resistant Whatman cellulose acetate membranes can filter hot liquids and hot gases.
Features and Benefits
- Minimal sample loss in protein-based samples due to low protein binding
- Pure cellulose acetate composition, minimizing sample contamination
- Hydrophilic, for use with aqueous and alcoholic media
- High solvent resistance, particularly to low molecular weight alcohols
- High physical strength and a heat resistance up to 180°C
- Can be sterilized by most common methods without sacrificing membrane integrity
Other Notes
Legal Information
Sold under license to DE10102744 and foreign equivalents thereof
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service