Documents
WHA10308210
Whatman® filter paper, Grade 989
circles, diam. 110 mm, pack of 100
Synonym(s):
Z749028
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
circles
packaging
pack of 100
manufacturer/tradename
Whatman 10308210
Whatman Article No. 28414152 (US reference)
parameter
medium flow rate
diam.
110 mm
pore size
8-10 μm
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva