W6853
Fumaric acid-1,4-13C2,2,3-d2
API for Clinical Studies, ≥98 atom % D, ≥99 atom % 13C, ≥98% (CP)
Synonym(s):
Fumaric acid-1,4-13C2,2,3-d2
About This Item
Recommended Products
grade
cGMP
isotopic purity
≥99 atom % 13C
≥98 atom % D
Assay
≥98% (CP)
greener alternative product score
old score: 59
new score: 48
Find out more about DOZN™ Scoring
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
mp
298-300 °C (subl.) (lit.)
greener alternative category
mass shift
M+4
storage temp.
15-25°C
SMILES string
O[13C](/C([2H])=C([2H])/[13C](O)=O)=O
Looking for similar products? Visit Product Comparison Guide
General description
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service