Skip to Content
Merck
All Photos(1)

Documents

W376302

Sigma-Aldrich

Dihydrojasmone

≥98%, stabilized, FG

Synonym(s):

3-Methyl-2-pentyl-2-cyclopenten-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C5H4(CH3)(=O)
CAS Number:
Molecular Weight:
166.26
FEMA Number:
3763
Beilstein:
1906471
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.140
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

contains

α-tocopherol as stabilizer (synthetic)

refractive index

n20/D 1.479 (lit.)

bp

120-121 °C/12 mmHg (lit.)

density

0.916 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; jasmine; floral; fruity

SMILES string

CCCCCC1=C(C)CCC1=O

InChI

1S/C11H18O/c1-3-4-5-6-10-9(2)7-8-11(10)12/h3-8H2,1-2H3

InChI key

YCIXWYOBMVNGTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service