Skip to Content
Merck
All Photos(1)

Key Documents

W361208

Sigma-Aldrich

1-Octen-3-yl butyrate

≥97%, stabilized, FCC, FG

Synonym(s):

Butanoic acid 1-octen-3-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
3612
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.282

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC

Assay

≥97%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.4295 (lit.)

bp

225-229 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

mushroom; fruity; buttery

SMILES string

CCCCCC(OC(=O)CCC)C=C

InChI

1S/C12H22O2/c1-4-7-8-10-11(6-3)14-12(13)9-5-2/h6,11H,3-5,7-10H2,1-2H3

InChI key

LZWFXVJBIZIHCH-UHFFFAOYSA-N

General description

Natural occurrence: Passion fruit.

Biochem/physiol Actions

Odor at 1.0%
Taste at 5 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

209.8 °F - closed cup

Flash Point(C)

98.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 58-58 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service