Skip to Content
Merck
All Photos(1)

Documents

W341606

Sigma-Aldrich

trans-3-Octen-2-one

≥98%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CHCOCH3
CAS Number:
Molecular Weight:
126.20
FEMA Number:
3416
EC Number:
Council of Europe no.:
11170
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.107
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

contains

α-tocopherol as stabilizer (synthetic)

refractive index

n20/D 1.448 (lit.)

bp

100 °C/18 mmHg (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

mushroom; earthy; spicy; herbaceous; sweet

SMILES string

[H]\C(CCCC)=C(\[H])C(C)=O

InChI

1S/C8H14O/c1-3-4-5-6-7-8(2)9/h6-7H,3-5H2,1-2H3/b7-6+

InChI key

ZCFOBLITZWHNNC-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service