Skip to Content
Merck
All Photos(1)

Key Documents

W340308

Sigma-Aldrich

cis-3-Hexenyl hexanoate

FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
198.30
FEMA Number:
3403
EC Number:
Council of Europe no.:
11779
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.271
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.473 (lit.)

bp

115 °C/15 mmHg (lit.)

density

0.88 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; fruity; waxy

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)CCCCC

InChI

1S/C12H22O2/c1-3-5-7-9-11-14-12(13)10-8-6-4-2/h5,7H,3-4,6,8-11H2,1-2H3/b7-5-

InChI key

RGACQXBDYBCJCY-ALCCZGGFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Can be used to modify cis-3-Hexenyl acetate or as part of fruity-green topnote complexes in fragrance applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fida Hussain Magsi et al.
Environmental entomology, 50(3), 589-598 (2021-03-08)
Tea black tussock moth, Dasychira baibarana (Matsumura) (Lepidoptera: Lymantriidae), is a devastating pest species of the tea plant in China. Here, we evaluated the responses of D. baibarana to tea plant volatiles using gas chromatography coupled electroantennographic detection (GC-EAD), eleclectroantennography
Yuting Yan et al.
Pesticide biochemistry and physiology, 165, 104542-104542 (2020-05-04)
The tea geometrid moth Ectropis obliqua Prout is one of the most serious moth pests in tea plants, and its sex pheromones have been identified as typical Type-II polyunsaturated hydrocarbons and epoxide derivatives. Therefore, the E. obliqua male olfactory system
Zhihui Feng et al.
Food research international (Ottawa, Ont.), 129, 108841-108841 (2020-02-11)
The orchid-like odor of a tea infusion is regarded as a noble aroma and an essential sensory attribute for certain premium teas. Such tea leaves are difficult to make and the quality is not always reproducible. This study is focused
Yaoyao Peng et al.
Molecules (Basel, Switzerland), 24(11) (2019-05-31)
Feijoa is an aromatic fruit and the essential oil from feijoa peel could be a valuable by-product in the juicing industry. An initial comparison of the essential oil extraction methods, steam-distillation and hydro-distillation, was conducted. The volatile compounds in the
Yaoyao Peng et al.
Food research international (Ottawa, Ont.), 129, 108873-108873 (2020-02-11)
The phenolic compounds and aroma active compounds in feijoa (Acca sellowiana (O.Berg) Burret) juice from four New Zealand grown cultivars (Apollo, Unique, Opal Star, and Wiki Tu) were investigated. A high total phenolic content (maximum 1.89 mg GAE/mL juice) and significant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service