Skip to Content
Merck
All Photos(1)

Key Documents

W305502

Sigma-Aldrich

Tetrahydrofurfuryl acetate

≥97%, FG

Synonym(s):

Tetrahydro-2-furylmethyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

W305502-SAMPLE-K
372,00 kr.
1 KG
1.320,00 kr.

372,00 kr.


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W305502-SAMPLE-K
372,00 kr.
1 KG
1.320,00 kr.

About This Item

Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17
FEMA Number:
3055
EC Number:
Council of Europe no.:
2069
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.166
NACRES:
NA.21
Organoleptic:
caramel; coffee; brown; pungent
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

372,00 kr.


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.437 (lit.)

bp

194-195 °C/753 mmHg (lit.)

density

1.061 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; coffee; brown; pungent

SMILES string

CC(=O)OCC1CCCO1

InChI

1S/C7H12O3/c1-6(8)10-5-7-3-2-4-9-7/h7H,2-5H2,1H3

InChI key

AAQDYYFAFXGBFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service