Skip to Content
Merck
All Photos(1)

Documents

W276804

Sigma-Aldrich

Nerolin bromelia

≥99%, FG

Synonym(s):

2-Ethoxynaphthalene, 2-naphthol ethyl ether, Ethyl 2-naphthyl ether, Neroline, beta-naphthol ethyl ether, beta-naphthyl ethyl ether, bromelia, ethyl beta-naphtholate, ethyl beta-naphthyl ether, nerolin bromelia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12O
CAS Number:
Molecular Weight:
172.22
FEMA Number:
2768
Beilstein:
2042397
EC Number:
Council of Europe no.:
2058
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.033
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets requirements for JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥99%

origin

China origin

mp

35-37 °C (lit.)

solubility

alcohol: soluble
dipropylene glycol: soluble
isopropyl myristate: soluble

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; citrus

SMILES string

CCOc1ccc2ccccc2c1

InChI

1S/C12H12O/c1-2-13-12-8-7-10-5-3-4-6-11(10)9-12/h3-9H,2H2,1H3

InChI key

GUMOJENFFHZAFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nerolin bromelia is an alkyl ether of 2-naphthol, which is generally used as a perfume and flavoring compound.

Application

The fluorescence of 2-Ethoxynaphthalene has been shown to be weakly quenched by m-dimethlphthalate and methyl benzoate. Importantly, at increased concentration methyl benzoate enhances the fluorescence by increasing the polarity of the solvent .

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of zeolite catalysts for etherification reaction
Kim JW, et al.
Catalysis Today, 87(1-4), 195-203 (2003)
Etherification of β-naphthol with alkanols using modified clays and sulfated zirconia
Yadav GD and Krishnan MS
Industrial & Engineering Chemistry Research, 37(8), 3358-3365 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service