W274100
Methyl 3-phenylpropionate
≥99%
Synonym(s):
Methyl hydrocinnamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH2CH2CO2CH3
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2741
EC Number:
Council of Europe no.:
427
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.746
Recommended Products
grade
Kosher
Assay
≥99%
refractive index
n20/D 1.502 (lit.)
bp
91-92 °C/4 mmHg (lit.)
density
1.043 g/mL at 25 °C (lit.)
SMILES string
COC(=O)CCc1ccccc1
Biochem/physiol Actions
Taste at 5 ppm
Other Notes
Natural occurrence: Tabasco peppers.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
212.0 °F - closed cup
Flash Point(C)
100.00 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mosciano, G.
Perfumer & Flavorist, 31, 53-53 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service