Skip to Content
Merck
All Photos(1)

Key Documents

W205818

Sigma-Aldrich

Isoamyl benzoate

≥98%, FCC, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W205818-SAMPLE-K
372,00 kr.
1 KG
765,00 kr.
10 KG
3.660,00 kr.

372,00 kr.


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W205818-SAMPLE-K
372,00 kr.
1 KG
765,00 kr.
10 KG
3.660,00 kr.

About This Item

Linear Formula:
C6H5CO2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
192.25
FEMA Number:
2058
EC Number:
Council of Europe no.:
562
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.755
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

372,00 kr.


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.494 (lit.)

bp

261-262 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; fruity; waxy; sweet

SMILES string

CC(C)CCOC(=O)c1ccccc1

InChI

1S/C12H16O2/c1-10(2)8-9-14-12(13)11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3

InChI key

MLLAPOCBLWUFAP-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aymerick Eudes et al.
Microbial cell factories, 15(1), 198-198 (2016-11-23)
BAHD acyltransferases, named after the first four biochemically characterized enzymes of the group, are plant-specific enzymes that catalyze the transfer of coenzyme A-activated donors onto various acceptor molecules. They are responsible for the synthesis in plants of a myriad of
Ki Yong Kim et al.
Environmental pollution (Barking, Essex : 1987), 227, 587-595 (2017-05-21)
Seasonal variations in the concentrations of eight ultraviolet (UV) filters and 22 illicit drugs including their metabolites in the Korean aquatic environment were investigated. Seawater samples from three beaches, water samples from two rivers, and influents and effluents from three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service