Skip to Content
Merck
All Photos(1)

Key Documents

T72400

Sigma-Aldrich

2′,4′,6′-Trimethylacetophenone

98%

Synonym(s):

2-Acetylmesitylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2COCH3
CAS Number:
Molecular Weight:
162.23
Beilstein:
2044595
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.517 (lit.)

bp

235-236 °C (lit.)
61-62 °C/0.1 mmHg (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1c(C)cc(C)cc1C

InChI

1S/C11H14O/c1-7-5-8(2)11(10(4)12)9(3)6-7/h5-6H,1-4H3

InChI key

XWCIICLTKWRWCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service