Skip to Content
Merck
All Photos(1)

Key Documents

SPC00024

Sigma-Aldrich

tert-Butyl 3-(hydroxymethyl)azetidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

OCC1CN(C(OC(C)(C)C)=O)C1

InChI

1S/C9H17NO3/c1-9(2,3)13-8(12)10-4-7(5-10)6-11/h7,11H,4-6H2,1-3H3

InChI key

HXRDRJKAEYHOBB-UHFFFAOYSA-N

Other Notes

Product of SpiroChem

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service