Skip to Content
Merck
All Photos(1)

Key Documents

QBD11026

Sigma-Aldrich

m-dPEG®24-DSPE

>95% (HPLC)

Synonym(s):

m-PEG24-DSPE, DSPE-PEG24-OMe, mPEG-DSPE, mPEG1000-DSPE, mPEG24-DSPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C91H180NO33P
Molecular Weight:
1847.36
MDL number:
UNSPSC Code:
12352200

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

m-dPEG24-DSPE is the lipid 1,2-Distearoyl-sn-glycero-3-phosphoethanolamine (DSPE) modified with a methoxy-terminated, single molecular weight, discrete polyethylene glycol (dPEG). This product is designed to protect liposomes and micelles from opsonization and elimination by the reticuloendothelial system (RES). The hydrophilic head is 74 atoms (86.2 Å) long and has a molecular weight of approximately 1100 Daltons. The dPEG24 is a monodisperse equivalent of the polymeric PEG1000.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jared F Stefanick et al.
ACS nano, 7(4), 2935-2947 (2013-02-21)
PEGylated liposomes are attractive pharmaceutical nanocarriers; however, literature reports of ligand-targeted nanoparticles have not consistently shown successful results. Here, we employed a multifaceted synthetic strategy to prepare peptide-targeted liposomal nanoparticles with high purity, reproducibility, and precisely controlled stoichiometry of functionalities
Gavin T Noble et al.
Trends in biotechnology, 32(1), 32-45 (2013-11-12)
Nanomedicine, particularly liposomal drug delivery, has expanded considerably over the past few decades, and several liposomal drugs are already providing improved clinical outcomes. Liposomes have now progressed beyond simple, inert drug carriers and can be designed to be highly responsive
Jared F Stefanick et al.
ACS nano, 7(9), 8115-8127 (2013-09-06)
Ligand-targeted nanoparticles are emerging drug delivery vehicles for cancer therapy. Here, we demonstrate that the cellular uptake of peptide-targeted liposomes and micelles can be significantly enhanced by increasing the hydrophilicity of the targeting peptide sequence while simultaneously optimizing the EG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service