Skip to Content
Merck
All Photos(1)

Key Documents

QBD10011

Sigma-Aldrich

Phthalimidooxy-dPEG®4-NHS ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
976,00 kr.
1000 MG
7.460,00 kr.

976,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
976,00 kr.
1000 MG
7.460,00 kr.

About This Item

Empirical Formula (Hill Notation):
C23H28N2O11
Molecular Weight:
508.48
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

976,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reactivity: amine reactive
reagent type: cross-linking reagent
reactivity: thiol reactive

functional group

NHS ester

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(ON1C(CCC1=O)=O)CCOCCOCCOCCOCCON2C(C(C=CC=C3)=C3C2=O)=O

InChI

1S/C23H28N2O11/c26-19-5-6-20(27)24(19)36-21(28)7-8-31-9-10-32-11-12-33-13-14-34-15-16-35-25-22(29)17-3-1-2-4-18(17)23(25)30/h1-4H,5-16H2

InChI key

HLAGLPXQSIDIDX-UHFFFAOYSA-N

Features and Benefits

Phthalimidooxy-dPEG®4-NHS ester allows introduction of an oxyamine function onto a target molecule for cross-linking proteins or for creating stable conjugates with the target molecule. The oxyamine function is protected by a phthalimido protecting group on one end of the spacer arm, and the spacer is installed on the target molecule with the amine-reactive N-hydroxysuccinimide (NHS) group on the other end of the spacer. The single molecular weight dPEG® spacers have precise length for spatial control. They are hydrophilic and non-immunogenic. The dPEG® spacers improve water solubility and reduce aggregation and precipitation of proteins. The phthalimido protecting group can be removed using hydrazine. The oxyamine group exposed after deprotection can be reacted with an aldehyde- or ketone-containing molecule to form an oxime bond linking the two molecules. These reagents can be used with any protein, peptide, or small molecule that has a free amine.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service