Skip to Content
Merck
All Photos(1)

Key Documents

O9301

Sigma-Aldrich

Oxamic acid hydrazide

99%

Synonym(s):

‘Semioxamazide’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COCONHNH2
CAS Number:
Molecular Weight:
103.08
Beilstein:
1751846
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

mp

216-218 °C (dec.) (lit.)

SMILES string

NNC(=O)C(N)=O

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for aldehydes and ketones.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service