Key Documents
MT51343192
Mettler Toledo InLab® Reference Flow electrode
Reference electrode with triple ceramic junction
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
material
glass shaft
description
S7 connector
Without cable
manufacturer/tradename
Mettler-Toledo 51343192
temperature measuring range
0-130 °C
L × W
120 mm × 12 mm
General description
Mettler Toledo InLab® Reference Flow is a glass-body, reference electrode with S7 screw head. Filled with 3 mol/L KCl for routine applications
  • Triple ceramic diaphragm
  • Electrolyte refill hose coupling
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)