Skip to Content
Merck
All Photos(3)

Documents

M38605

Sigma-Aldrich

3-Methylcyclohexanone

97%

Synonym(s):

Tetrahydro-m-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H9(=O)
CAS Number:
Molecular Weight:
112.17
Beilstein:
969804
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

169-170 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

SMILES string

CC1CCCC(=O)C1

InChI

1S/C7H12O/c1-6-3-2-4-7(8)5-6/h6H,2-5H2,1H3

InChI key

UJBOOUHRTQVGRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service